FRO026591363 Frontier Scientific Services Inc.

O-Acetylphenyl Benzoate

  • MDL number MFCD00017235
  • Cas Number : 4010-33-7
  • Molecular Weight : 240.2611

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Salt Data Free
Country of Origin USA
Availability details for
Quantity
  • Availability
  • Add to Cart