FRO026591368 Frontier Scientific Services Inc.

2-Bromononane

  • MDL number MFCD00017865
  • Cas Number : 2216-35-5
  • Molecular Weight : 207.1558

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Salt Data Free
Country of Origin USA
Availability details for
Quantity
  • Availability
  • Add to Cart