FRO026591394 Frontier Scientific Services Inc.

1-Chloro-3,3-DiMethyl-Butane

  • MDL number MFCD00018980
  • Cas Number : 2855-08-5
  • Molecular Weight : 120.620003

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Salt Data Free
Country of Origin USA
Availability details for
Quantity
  • Availability
  • Add to Cart