FRO026591403 Frontier Scientific Services Inc.

1-Chloro-4-(4-Methylphenoxy)Butane

  • MDL number MFCD00019004
  • Cas Number : 71720-44-0
  • Molecular Weight : 198.6946

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Salt Data Free
Country of Origin USA
Availability details for
Quantity
  • Availability
  • Add to Cart