FRO026591453 Frontier Scientific Services Inc.

2-(Pyrrolidin-1-Yl-Methyl)Pyridin-3-Ol

  • MDL number MFCD00020846
  • Cas Number : 7149-45-3
  • Molecular Weight : 178.231003

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Salt Data Free
Country of Origin USA
Availability details for
Quantity
  • Availability
  • Add to Cart