FRO026591538 Frontier Scientific Services Inc.

N4-Succinoylsulfanilamide

  • MDL number MFCD00025390
  • Cas Number : 3563-14-2
  • Molecular Weight : 272.28154

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Salt Data Free
Country of Origin USA
Availability details for
Quantity
  • Availability
  • Add to Cart