FRO026591559 Frontier Scientific Services Inc.

N-(4-Methoxybenzyl)Acetoacetamide

  • MDL number MFCD00026261
  • Molecular Weight : 221.2583

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Salt Data Free
Country of Origin USA
Availability details for
Quantity
  • Availability
  • Add to Cart