FRO026591560 Frontier Scientific Services Inc.

N-(3-PHENYLPROPYL)ACETOACETAMIDE

  • MDL number MFCD00026264
  • Molecular Weight : 219.28594

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Salt Data FREE
Country of Origin USA
Availability details for
Quantity
  • Availability
  • Add to Cart