FRO026591774 Frontier Scientific Services Inc.

2-[(Diethylamino)Methyl]Pyridin-3-Ol

  • MDL number MFCD00085424
  • Cas Number : 2168-14-1
  • Molecular Weight : 180.246796

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Salt Data Free
Country of Origin USA
Availability details for
Quantity
  • Availability
  • Add to Cart