FRO026591792 Frontier Scientific Services Inc.

2-ACETOACETAMIDO-3-METHYLPYRIDINE

  • MDL number MFCD00085948
  • Molecular Weight : 192.2193

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Salt Data FREE
Country of Origin USA
Availability details for
Quantity
  • Availability
  • Add to Cart