FRO026591794 Frontier Scientific Services Inc.

1-Acetoacetyl-2-Methylpiperidine

  • MDL number MFCD00085950
  • Molecular Weight : 183.2525

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Salt Data Free
Country of Origin USA
Availability details for
Quantity
  • Availability
  • Add to Cart