FRO026592063 Frontier Scientific Services Inc.

2-(4-Fluorophenyl)-2-Phenylethylamine Hydrochloride

  • MDL number MFCD02089463
  • Molecular Weight : 251.73

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Salt Data Hydrochloride
Country of Origin USA
Availability details for
Quantity
  • Availability
  • Add to Cart