FRO026592341 Frontier Scientific Services Inc.

2-[(4-Methylphenyl)Thio]Pyridine-3-Carbonyl Chloride

  • MDL number MFCD00067835
  • Cas Number : 175135-78-1
  • Molecular Weight : 263.74775

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Salt Data Free
Country of Origin USA
Availability details for
Quantity
  • Availability
  • Add to Cart