FRO026592385 Frontier Scientific Services Inc.

1-(5-Bromo-1-Benzofuran-2-Yl)Ethan-1-One

  • MDL number MFCD00276737
  • Cas Number : 38220-75-6
  • Molecular Weight : 239.065002

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Salt Data Free
Country of Origin USA
Availability details for
Quantity
  • Availability
  • Add to Cart