FRO026592390 Frontier Scientific Services Inc.

N-[(2-Bromophenyl)Methyl]Acetamide

  • MDL number MFCD00831169
  • Cas Number : 74315-07-4
  • Molecular Weight : 228.085999

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Salt Data Free
Country of Origin USA
Availability details for
Quantity
  • Availability
  • Add to Cart