FRO026592402 Frontier Scientific Services Inc.

1-(3,5-Dichlorophenyl)-1H-Pyrrole-2-Carbaldehyde

  • MDL number MFCD00174281
  • Cas Number : 175136-79-5
  • Molecular Weight : 240.085007

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Salt Data Free
Country of Origin USA
Availability details for
Quantity
  • Availability
  • Add to Cart