FRO026592412 Frontier Scientific Services Inc.

1-Tert-Butyl-3,5-Dimethyl-1H-Pyrazole-4-Carbaldehyde

  • MDL number MFCD00120293
  • Cas Number : 647824-51-9
  • Molecular Weight : 180.246796

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Salt Data Free
Country of Origin USA
Availability details for
Quantity
  • Availability
  • Add to Cart