FRO026592481 Frontier Scientific Services Inc.

4-N-Trifluoroacetamidophenol

  • MDL number MFCD00270089
  • Molecular Weight : 205.13772

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Salt Data Free
Country of Origin USA
Availability details for
Quantity
  • Availability
  • Add to Cart