FRO026592610 Frontier Scientific Services Inc.

2-(2H-Tetrazol-5-Yl)-Pyrazine

  • MDL number MFCD01569289
  • Molecular Weight : 148.1278

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Salt Data Free
Country of Origin USA
Availability details for
Quantity
  • Availability
  • Add to Cart