FRO026592721 Frontier Scientific Services Inc.

3-Phenyl-5-(Trifluoromethyl)-1H-Pyrazole

  • MDL number MFCD00764002
  • Molecular Weight : 212.171204

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Salt Data Free
Country of Origin USA
Availability details for
Quantity
  • Availability
  • Add to Cart