FRO026592746 Frontier Scientific Services Inc.

N'-Hydroxy-2,5-Dimethoxybenzenecarboximidamide

  • MDL number MFCD05864457
  • Molecular Weight : 196.2076

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Salt Data Free
Country of Origin USA
Availability details for
Quantity
  • Availability
  • Add to Cart