FRO026592749 Frontier Scientific Services Inc.

2-(2H-1,2,3,4-Tetraazol-5-Yl-Methyl)-1,3-Benzothiazole

  • MDL number MFCD00206465
  • Molecular Weight : 217.2536

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Salt Data Free
Country of Origin USA
Availability details for
Quantity
  • Availability
  • Add to Cart