FRO026592765 Frontier Scientific Services Inc.

5-Cyclopropyl-1H-Pyrazole-4-Carbonitrile

  • MDL number MFCD00219866
  • Molecular Weight : 133.1533

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Salt Data Free
Country of Origin USA
Availability details for
Quantity
  • Availability
  • Add to Cart