FRO026592770 Frontier Scientific Services Inc.

2-(1-Benzothiophen-3-Yl)-N'-Hydroxyethanimidamide

  • MDL number MFCD03791156
  • Molecular Weight : 206.268

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Salt Data Free
Country of Origin USA
Availability details for
Quantity
  • Availability
  • Add to Cart