FRO026592788 Frontier Scientific Services Inc.

2-[4-(1,3,4-Oxadiazol-2-Yl)Phenyl]Acetonitrile

  • MDL number MFCD01934464
  • Molecular Weight : 185.182098

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Salt Data Free
Country of Origin USA
Availability details for
Quantity
  • Availability
  • Add to Cart