FRO026592844 Frontier Scientific Services Inc.

N-Hydroxy-2-(4-Methoxy-Phenoxy)-Acetamidine

  • MDL number MFCD00245494
  • Molecular Weight : 196.2048

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Salt Data Free
Country of Origin USA
Availability details for
Quantity
  • Availability
  • Add to Cart