FRO026592854 Frontier Scientific Services Inc.

3-(Tert-Butyl)Pyrazole-4-Carbonitrile

  • MDL number MFCD00121111
  • Molecular Weight : 149.197

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Salt Data Free
Country of Origin USA
Availability details for
Quantity
  • Availability
  • Add to Cart