FRO026593020 Frontier Scientific Services Inc.

6-(2,4-Dichlorophenoxy)Pyridin-3-Amine

  • MDL number MFCD00052643
  • Cas Number : 85686-48-2
  • Molecular Weight : 255.100006

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Salt Data Free
Country of Origin USA
Availability details for
Quantity
  • Availability
  • Add to Cart