FRO026593094 Frontier Scientific Services Inc.

2-(4-Carbamoylphenoxy)-2-Methylpropionic Acid

  • MDL number MFCD00084921
  • Cas Number : 121809-54-9
  • Molecular Weight : 223.2306

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Salt Data Free
Country of Origin USA
Availability details for
Quantity
  • Availability
  • Add to Cart