FRO026593096 Frontier Scientific Services Inc.

4,4,4-Trichlorobutanoic Acid

  • MDL number MFCD00100736
  • Cas Number : 2345-32-6
  • Molecular Weight : 191.440002

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Salt Data Free
Country of Origin USA
Availability details for
Quantity
  • Availability
  • Add to Cart