FRO026593188 Frontier Scientific Services Inc.

2-CHLORO-5-[2-(3-CYANOPHENYL)-1-OXOETHYL]PYRIDINE

  • MDL number MFCD02260450
  • Molecular Weight : 256.69363

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Salt Data FREE
Country of Origin USA
Availability details for
Quantity
  • Availability
  • Add to Cart