FRO026593300 Frontier Scientific Services Inc.

2-(2,4-Dichlorophenylthio)Acetic Acid

  • MDL number MFCD00052119
  • Molecular Weight : 237.1058

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Salt Data Free
Country of Origin USA
Availability details for
Quantity
  • Availability
  • Add to Cart