FRO026593631 Frontier Scientific Services Inc.

1,1,1-Trifluoro-3-Phenylpropan-2-Ol

  • MDL number MFCD00178722
  • Molecular Weight : 190.162399

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Salt Data Free
Country of Origin USA
Availability details for
Quantity
  • Availability
  • Add to Cart