FRO026593795 Frontier Scientific Services Inc.

2-(Acetylamino)-3-[4-(Trifluoromethoxy)Phenyl]Propanoic Acid

  • MDL number MFCD01313663
  • Molecular Weight : 291.22894

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Salt Data Free
Country of Origin USA
Availability details for
Quantity
  • Availability
  • Add to Cart