FRO026594174 Frontier Scientific Services Inc.

2,4-Dimethoxyphenyl Isothiocyanate

  • MDL number MFCD00041058
  • Cas Number : 33904-03-9
  • Molecular Weight : 195.2416

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Salt Data Free
Country of Origin USA
Availability details for
Quantity
  • Availability
  • Add to Cart