FRO048767309 Frontier Scientific Services Inc.

GLYCINE N-PENTYL ESTER HYDROCHLORIDE

  • MDL number MFCD00151828
  • Molecular Weight : 181.6631

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Salt Data HYDROCHLORIDE
Country of Origin USA
Availability details for
Quantity
  • Availability
  • Add to Cart