FRO048767332 Frontier Scientific Services Inc.

1-Ethyl-1H,2H,3H,4H-Pyrrolo[1,2-A]Pyrazine Hydrochloride

  • MDL number MFCD07781050
  • Molecular Weight : 186.681901

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Salt Data Hydrochloride
Country of Origin USA
Availability details for
Quantity
  • Availability
  • Add to Cart