FRO048767363 Frontier Scientific Services Inc.

C-(6-Chloro-1H-Benzoimidazol-2-Yl)-Methylamine Hydrochloride

  • MDL number MFCD09038197
  • Cas Number : 273399-95-4
  • Molecular Weight : 218.082994

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Salt Data Hydrochloride
Country of Origin USA
Availability details for
Quantity
  • Availability
  • Add to Cart