FRO048767401 Frontier Scientific Services Inc.

L-2-Aminopentanedioic Acid Sodium Salt

  • MDL number MFCD00150138
  • Cas Number : 6106-04-3
  • Molecular Weight : 187.126

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Salt Data Sodium Salt
Country of Origin USA
Availability details for
Quantity
  • Availability
  • Add to Cart