FRO048767403 Frontier Scientific Services Inc.

2'-BenzoYl-Oxy-6'-Methoxyacetophenone

  • MDL number MFCD00017239
  • Molecular Weight : 270.2876

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Salt Data Free
Country of Origin USA
Availability details for
Quantity
  • Availability
  • Add to Cart