FRO048767579 Frontier Scientific Services Inc.

2-(1H-Imidazol-4-Yl)Acetic Acid Hydrochloride

  • MDL number MFCD00012698
  • Cas Number : 3251-69-2
  • Molecular Weight : 162.574297

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Salt Data Hydrochloride
Country of Origin USA
Availability details for
Quantity
  • Availability
  • Add to Cart