FRO048767628 Frontier Scientific Services Inc.

3-Chloroquinuclidine Hydrochloride

  • MDL number MFCD00012728
  • Cas Number : 33601-77-3
  • Molecular Weight : 182.0934

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Salt Data Hydrochloride
Country of Origin USA
Availability details for
Quantity
  • Availability
  • Add to Cart