FRO048767687 Frontier Scientific Services Inc.

4-Thiocyanatophenylhydrazine Hydrochloride

  • MDL number MFCD00035459
  • Cas Number : 74411-22-6
  • Molecular Weight : 201.68

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Salt Data Hydrochloride
Country of Origin USA
Availability details for
Quantity
  • Availability
  • Add to Cart