FRO048767689 Frontier Scientific Services Inc.

6-Amino-2,4-Dichloro-3-Methylphenol Hydrochloride

  • MDL number MFCD00035472
  • Cas Number : 40677-44-9
  • Molecular Weight : 228.5059

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Salt Data Hydrochloride
Country of Origin USA
Availability details for
Quantity
  • Availability
  • Add to Cart