FRO048767700 Frontier Scientific Services Inc.

3-Amino-4-Mercaptobenzotrifluoride Hydrochloride

  • MDL number MFCD00042600
  • Cas Number : 4274-38-8
  • Molecular Weight : 229.6518

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Salt Data Hydrochloride
Country of Origin USA
Availability details for
Quantity
  • Availability
  • Add to Cart