FRO048767709 Frontier Scientific Services Inc.

3-Aminoquinuclidine Dihydrochloride Dihydrochloride

  • MDL number MFCD00137395
  • Cas Number : 6530-09-2
  • Molecular Weight : 199.123

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Salt Data Dihydrochloride
Country of Origin USA
Availability details for
Quantity
  • Availability
  • Add to Cart