FRO048767717 Frontier Scientific Services Inc.

Cis-4-Chloro-2-Butenylamine Hydrochloride

  • MDL number MFCD00239421
  • Cas Number : 7153-66-4
  • Molecular Weight : 142.03

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Salt Data Hydrochloride
Country of Origin USA
Availability details for
Quantity
  • Availability
  • Add to Cart