FRO048767784 Frontier Scientific Services Inc.

4-(4-Bromophenyl)-2-Methylpyrimidine

  • MDL number MFCD02089754
  • Cas Number : 124276-67-1
  • Molecular Weight : 249.106995

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Salt Data Free
Country of Origin USA
Availability details for
Quantity
  • Availability
  • Add to Cart