FRO048767890 Frontier Scientific Services Inc.

Methyl[(3-Phenyl-1,2,4-Oxadiazol-5-Yl)Methyl]Amine Hydrochloride

  • MDL number MFCD07781051
  • Molecular Weight : 225.674806

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Salt Data Hydrochloride
Country of Origin USA
Availability details for
Quantity
  • Availability
  • Add to Cart