FRO048767938 Frontier Scientific Services Inc.

Methyl[(4-Methyl-1,3-Thiazol-2-Yl)Methyl]Amine Dihydrochloride

  • MDL number MFCD08062597
  • Molecular Weight : 215.144

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Salt Data Dihydrochloride
Country of Origin USA
Availability details for
Quantity
  • Availability
  • Add to Cart